missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Bat IgG (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB7237
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Bat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Bat | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:30000, Flow Cytometry, ELISA 1:1000 - 1:30000, Immunohistochemistry 1:200- 1:2000, Immunocytochemistry/Immunofluorescence 1:200- 1:2000, Immunohistochemistry-Paraffin 1:200-1:2000 | |
| Bat IgG-heavy and light chain | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with Bat IgG and with light chains common to other Bat immunoglobulins. Antibody has been shown to react with bat genus species Pteropus vampirus, Desmodus rotundus, Eptesicus fuscus, Tadrida pumila, T. condylura, Hypsignathus monstrosus, Rosettus aegyptiacus, Epomorphus crypturus, Molossus species. And Phylostomus species. No antibody was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction