missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Armenian Hamster IgG (H+L) Secondary Antibody, DyLight 350, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Armenian Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot, ChIP assay (ChIP)
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot, ChIP Assay |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Host Species | Goat |
| Immunogen | Armenian Hamster IgG whole molecule |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?