missing translation for 'onlineSavingsMsg'
Learn More

Goat Anti-Armenian hamster IgG H&L (Alkaline Phosphatase), Abcam

Product Code. 32114333
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32114333 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32114333 Supplier Abcam Supplier No. ab5746500ug
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Goat Anti-Armenian hamster IgG H&L (Alkaline Phosphatase)

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Armenian hamster IgG
Applications Dot Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation pH: 8, Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 1% BSA, 0.88% Sodium chloride, 0.788% Tris HCl, 0.00952% Magnesium chloride, 0.00136% Zinc chloride
Host Species Goat
Immunogen Armenian hamster IgG
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Armenian Hamster
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Molecule Armenian hamster IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.