missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anit-Rabbit IgG, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals
Brand: MP Biomedicals 0855689
This item is not returnable.
View return policy
Description
Horseradish peroxidase (HRP)-conjugated goat affinity purified antibody to rabbit IgG (whole molecule) and buffer salts.
Host Species: Goat
Species Reactivity: Rabbit
Specifications
| IgG, Whole Molecule | |
| HRP | |
| 2 mL | |
| Rabbit |
| Polyclonal | |
| Goat | |
| Secondary | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction