missing translation for 'onlineSavingsMsg'
Learn More

GNPAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16863854 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16863854 20 μL 20µL
16853854 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16863854 Supplier Proteintech Supplier No. 149311AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody recognize the 69 kDa GNPAT protein.

GNPAT is a key enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in GNPAT are associated with rhizomelic chondrodysplasia punctata type 2, which is characterized by rhizomelic shortening of femur and humerus, vertebral disorders, cataract, cutaneous lesions and severe mental retardation.

Specifications

Antigen GNPAT
Applications Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GNPAT
Gene Accession No. O15228, P98192, Q9ES71
Gene Alias DAP AT, DAPAT, DHAP AT, DHAPAT, GNPAT
Gene Symbols GNPAT
Host Species Rabbit
Immunogen GNPAT Fusion Protein Ag6735
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14712, 8443, 84470
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.