missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody recognize the 69 kDa GNPAT protein.
GNPAT is a key enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in GNPAT are associated with rhizomelic chondrodysplasia punctata type 2, which is characterized by rhizomelic shortening of femur and humerus, vertebral disorders, cataract, cutaneous lesions and severe mental retardation.
Specifications
Specifications
| Antigen | GNPAT |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.19 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GNPAT |
| Gene Accession No. | O15228, P98192, Q9ES71 |
| Gene Alias | DAP AT, DAPAT, DHAP AT, DHAPAT, GNPAT |
| Gene Symbols | GNPAT |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?