missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ GNG3 (Human) Recombinant Protein

Human GNG3 full-length ORF ( AAH15563, 1 a.a. - 75 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00002785-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105581

  • 349.00€ / 10µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MKGETPVNSTMSIGQARKMVEQLKIEASLCRIKVSKAAADLVTYCDAHACEDPLITPVPTSENPFREKKFFCALL
Specifications

Specifications

AAH15563
guanine nucleotide binding protein (G protein), gamma 3
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
2785
Wheat germ expression system
10 μg
GNG3
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers