Learn More
GNB3 Mouse anti-Human, Unconjugated, Clone: 2B6E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67497-1-IG-20UL
Description
The GNB3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNB3. This antibody recognizes human, mouse, rat, pig antigen. The GNB3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| GNB3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2B6E1 | |
| Unconjugated | |
| 20 μL | |
| 2784 | |
| IgG1 |