missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ GNAI2 (Human) Recombinant Protein

Human GNAI2 full-length ORF ( AAH16995, 1 a.a. - 339 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00002771-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195541

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MEYAGHLPASSAQGTILACTSCTGAGESGKSTIVKQMKIIHEDGYSEEECRQYRAVVYSNTIQSIMAIVKAMGNLQIDFADPSRADDARQLFALSCTAEEQGVLPDDLSGVIRRLWADHGVQACFGRSREYQLNDSAAYYLNDLERIAQSDYIPTQQDVLRTRVKTTGIVETHFTFKDLHFKMFDVGGQRSERKKWIHCFEGVTAIIFCVALSAYDLVLAEDEEMNRMHESMKLFDSICNNKWFTDTSIILFLNKKDLFEEKITHSPLTICFPEYTGANKYDEAASYIQSKFEDLNKRKDTKEIYTHFTCATDTKNVQFVFDAVTDVIIKNNLKDCGLF
Specifications

Specifications

AAH16995
guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GNAI2
GST
2771
Wheat germ expression system
25 μg
GIP, GNAI2B, H_LUCA15.1, H_LUCA16.1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers