Learn More
Liebherr™ GN Series Freezer
Product Code. p-8002211 Shop All Liebherr Products
Click to view available options
Capacity (Metric):
307 L
Unit Size:
Each
Product Code. 12527066
Brand: Liebherr™ GN 3113
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these freezers for general laboratory storage of temperature-sensitive samples. Liebherr™ GN Series Freezers provide expert NoFrost refrigeration technology to ensure long-term freshness.
Automatic SuperFrost function
  • Makes freezing simple and saves energy
  • quickly reduces the temperature to -32 °C and thus creates the cold air required to preserve vital vitamins
  • As soon as the food is completely frozen, the automatic SuperFrost function switches back to normal mode after max. 65 hours, and thus assists in saving electricity
FrostSafe
  • With the FrostSafe, the extra-high and removable drawers are closed all round
  • Cold cannot dissipate so quickly when the appliance is opened
  • Transparent front of the compartments guarantees an optimum overview of the frozen items
VarioSpace
  • All freezers with NoFrost and SmartFrost feature drawers and intermediate glass shelves underneath them which can be conveniently removed
  • This results in VarioSpace; the practical system for extra storage space so that space can be created quickly even for larger frozen food items
Slimline door handle with integrated opening mechanism
  • The practical door handles with integrated opening mechanism ensure that the appliance doors can be opened with a minimum of effort even if they are opened several times in rapid succession
  • Handle mechanism assists in pushing the door from the appliance housing
Specifications
Capacity (Metric) 307 L
Temperature Range 10°C to 43°C
Color White
Model GN3113X
Dimensions (L x W x H) Exterior 155.5 H x 69.7 W x 75 D cm
Depth (Metric) Interior 48 cm
Height (Metric) Interior 122 cm
Width (Metric) Interior 48 cm
Voltage 230 V
Frequency 50 Hz
Refrigerant R600A