Learn More
Lewis Y Antibody (LWY/9243R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-23755-100ug
 View more versions of this product
Product Code. 18623755
  • 496.00€ / 100µg
Description
Lewis Y Monoclonal antibody specifically detects Lewis Y in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lewis Y
Monoclonal
0.2 mg/mL
Immunohistochemistry-Paraffin : 1-2 μg/mL
IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3
H3396 cultured line established from a metastasis of a human breast adenocarcinoma.
100 μg
Primary
Store at 4°C.
IgG κ
Immunohistochemistry (Paraffin)
LWY/9243R
Unconjugated
10 mM PBS with 0.05% BSA
Rabbit
Protein A or G purified