missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AcroBiosystems GMP Human TGF-Beta 1 / TGFB1 Protein
Description
This gene encodes a secreted ligand of the TGF-beta (transforming growth factor-beta) superfamily of proteins. Ligands of this family bind various TGF-beta receptors leading to recruitment and activation of SMAD family transcription factors that regulate gene expression. The encoded preproprotein is proteolytically processed to generate a latency-associated peptide (LAP) and a mature peptide, and is found in either a latent form composed of a mature peptide homodimer, a LAP homodimer, and a latent TGF-beta binding protein, or in an active form consisting solely of the mature peptide homodimer. The mature peptide may also form heterodimers with other TGFB family members. This encoded protein regulates cell proliferation, differentiation and growth, and can modulate expression and activation of other growth factors including interferon gamma and tumor necrosis factor alpha. This gene is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease. [provided by RefSeq, Aug 2016]
Specifications
Specifications
| Accession Number | Uniprot |
| Characterization | TGF-beta 1 Structure This protein carries no tag. The protein has a calculated MW of 12.8 kDa. The protein migrates as 13 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE). |
| Format | Powder-GMP |
| Formulation | Lyophilized from 0.22 μm filtered solution in 100 mM HAC, pH 3.0 with protectants. Contact us for customized product form or formulation. |
| Quality Control Testing | <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative |
| Quantity | 10 mg |
| Source | GMP Human TGF-Beta 1 Protein (GMP-TG1H25) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession # ). Predicted N-terminus: Ala 279 Request for sequence |
| Biological Activity | GMP Human TGF-Beta 1 Protein (Cat. No. GMP-TG1H25) inhibits the GMP Human IL-4 Protein (Cat. No. GMP-L04H26) dependent proliferation the TF-1 cells. The specific activity of GMP Human TGF-Beta 1 Protein is >6.00x10^6 IU/mg, which is calibrated against transforming growth factor β1 (NIBSC code:89/514) (QC tested).∣The activity of GMP Human TGF-Beta 1 Protein (Cat. No. GMP-TG1H25) was higher than other competing products.∣GMP Human FGF basic Protein (Cat. No. GMP-FGCH17) and GMP Human TGF-Beta 1 Protein (Cat. No. GMP-TG1H25) could maintain the stemness of hiPSCs with high expression of stem cell genes OCT4, SOX2, and SSEA4 with GMP Human Laminin 521 Protein (Cat. No. GMP-LA5H24).∣The Cell based assay shows that GMP Human TGF-Beta 1 Protein (Cat. No. GMP-TG1H25) is stable at 37°C for 24 hours.∣The Cell based assay shows that GMP Human TGF-Beta 1 Protein (Cat. No. GMP-TG1H25) is stable at 4°C for 180 days.∣The Cell based assay shows that GMP Human TGF-Beta 1 Protein (Cat. No. GMP-TG1H25) is stable after freezing and thawing 3 times.∣The Cell based assay shows batch-to-batch consistency between Acro's GMP and PG TGF-Beta 1. |
| Species | Human |
| Sterility | Sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?