missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM130/GOLGA2 Antibody (NN 2C10/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00€ - 453.00€
Specifications
| Antigen | GM130/GOLGA2 |
|---|---|
| Clone | NN 2C10/1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18492009
|
Novus Biologicals
NB100-2622-0.025mg |
0.025 mg |
190.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18591661
|
Novus Biologicals
NB100-2622 |
0.1 mg |
453.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GM130/GOLGA2 Monoclonal antibody specifically detects GM130/GOLGA2 in Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| GM130/GOLGA2 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Golgi Apparatus Markers | |
| PBS | |
| 2801 | |
| IgG2a κ | |
| Protein A purified |
| NN 2C10/1 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat | |
| 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
| Recombinant purified rat GM130/GOLGA2 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title