missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for ELISA, Sandwich ELISA.
Specifications
Specifications
| Antigen | GM-CSF |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Dilution | Sandwich ELISA |
| Gene Alias | colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim |
| Gene Symbols | CSF2 |
| Host Species | Mouse |
| Quantity | 15 Plates |
| Research Discipline | Biologically Active Proteins, Cytokine Research, Immunology |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?