missing translation for 'onlineSavingsMsg'
Learn More

Glypican 5 Antibody (297716), Allophycocyanin/Cy7, Novus Biologicals™

Product Code. 30033448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30033448 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30033448 Supplier Novus Biologicals Supplier No. FAB2607APCCY7

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glypican 5 Monoclonal antibody specifically detects Glypican 5 in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 5
Applications Flow Cytometry
Classification Monoclonal
Clone 297716
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias bA93M14.1, glypican 5, glypican proteoglycan 5, glypican-5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Glypican 5, Glu25-Thr554, Accession # P78333
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2262
Target Species Human, Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.