missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glypican 4 Monoclonal Antibody (965523)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562007
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. The GPC4 gene is adjacent to the 3' end of GPC3 and may also play a role in Simpson-Golabi-Behmel syndrome.
Specifications
| Glypican 4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O75487 | |
| GPC4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Glypican 4, Met1-Ser529. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA | |
| 965523 | |
| Unconjugated | |
| GPC4 | |
| 9530073D23Rik; AF311610; AI385680; AI661372; dJ900E8.1 (glypican 4); glypican 4; glypican proteoglycan 4; glypican-4; Gpc4; K-glypican; Secreted glypican-4; UNQ474/PRO937 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2239 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction