missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 1 Antibody (S03-2E8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19539-100ul
This item is not returnable.
View return policy
Description
Glypican 1 Monoclonal antibody specifically detects Glypican 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Glypican 1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2817 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| S03-2E8 | |
| Unconjugated | |
| FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1 | |
| A synthetic peptide of human Glypican 1/GPC1 (Uniprot # P35052) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction