missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-histidyl-L-lysine TFA Salt, TRC

Product Code. 30240985
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240985 250 mg 250mg
30240946 100 mg 100mg
30240762 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240985 Supplier Toronto Research Chemicals Supplier No. TRCG625240250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1846624-43-8
Molecular Formula C14H24N6O4 . C2HF3O2
Molecular Weight (g/mol) 454.4
IUPAC Name (2S)-6-amino-2-[[(2S)-2-[(2-aminoacetyl)amino]-3-(1H-imidazol-4-yl)propanoyl]amino]hexanoic acid;2,2,2-trifluoroacetic acid
SMILES NCCCC[C@H](NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)CN)C(=O)O.OC(=O)C(F)(F)F

Specifications

InChI Formula InChI=1S/C14H24N6O4.C2HF3O2/c15-4-2-1-3-10(14(23)24)20-13(22)11(19-12(21)6-16)5-9-7-17-8-18-9;3-2(4,5)1(6)7/h7-8,10-11H,1-6,15-16H2,(H,17,18)(H,19,21)(H,20,22)(H,23,24);(H,6,7)/t10-,11-;/m0./s1
Quantity 250 mg
Formula Weight 454.1788
Shipping Condition Room Temperature
Chemical Name or Material Glycyl-L-histidyl-L-lysine TFA Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.