CAS RN 1846624-43-8
CAS RN 1846624-43-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycyl-L-histidyl-L-lysine TFA Salt, TRC
CAS: 1846624-43-8 Molecular Formula: C14H24N6O4 . C2HF3O2 Molecular Weight (g/mol): 454.4 IUPAC Name: (2S)-6-amino-2-[[(2S)-2-[(2-aminoacetyl)amino]-3-(1H-imidazol-4-yl)propanoyl]amino]hexanoic acid;2,2,2-trifluoroacetic acid SMILES: NCCCC[C@H](NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)CN)C(=O)O.OC(=O)C(F)(F)F