missing translation for 'onlineSavingsMsg'
Learn More
Learn More
glycerol-3-phosphate permease Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69300
This item is not returnable.
View return policy
Description
glycerol-3-phosphate permease Polyclonal specifically detects glycerol-3-phosphate permease in Human samples. It is validated for Western Blot.
Specifications
| glycerol-3-phosphate permease | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ22340, G-3-P permease, G-3-P transporter, G3PPgene similar to glycerol-3-phosphate permease10Glycerol-3-phosphate permease, glycerol-3-phosphate transporter, solute carrier family 37 (glycerol-3-phosphate transporter), member 1, Solute carrier family 37 member 1 | |
| Rabbit | |
| 58 kDa | |
| 100 μL | |
| Primary | |
| Zebrafish: 79%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P57057 | |
| SLC37A1 | |
| Synthetic peptides corresponding to SLC37A1(solute carrier family 37 (glycerol-3-phosphate transporter), member 1) The peptide sequence was selected from the middle region of SLC37A1 (NP_061837). Peptide sequence LKIPGVIEFSLCLLFAKLVSYTFLFWLPLYITNVDHLDAKKAGELSTLFD The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 54020 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction