missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Glybenclamide (NLRP3 Inhibitor)

Product Code. 18133973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1000 mg
Unit Size:
1000mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18133973 1000 mg 1000mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18133973 Supplier Novus Biologicals™ Supplier No. NBP230141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glybenclamide, also known as glyburide, is an ATP-sensitive potassion channel inhibitor that has been shown to inhibit various biological processes including PAMP and DAMP-induced activation of the NRLP3 inflammasome as well as the activation of the inflammasome by crystalline substances

Form : White Solid Reconstitute: Dilute further with sterilized PBS. Make a stock solution of 25mg/mL of the NLRP3 inhibitor in high purity DMSO (>99.9%).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Inhibition of NLRP3
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1000 mg
Product Type Glybenclamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.