Learn More
Glutamine synthetase Mouse anti-Human, Porcine, Clone: 1D10G8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66323-1-IG-150UL
 View more versions of this product
Product Code. 16808095
  • 410.00€ / 150µL
Description
Structural maintece of chromosomes (SMC) and non-SMC condensin proteins associate into complexes that have been implicated in the process of chromosome condensation. A crucial prerequisite for accurate segregation of replicated sister chromatids is the condensation of the chromosomes into a manageable form prior to metaphase. The condensin I complex consists of two SMC subunits, SMC2 and SMC4, and three non-SMC subunits, CAP-H, CAP-G, and CAP-D2. An alternative complex, the condensin II complex, contains alternate non-SMC subunits, CAP-G2, CAP-H2, and CAP-D3. CAP-H2 is also known as Non-SMC condensin II complex, subunit H2 (NCAPH2) or kleisin beta isoform 2. The three non-SMC subunits in the condensing complexes form a regulatory subcomplex that is required to activate the SMC ATPases and to promote mitosis-specific chromatin binding of the holocomplex. The precise individual functions of each non-SMC protein in activation remain to be determined.
Specifications
Specifications
Glutamine synthetase
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15104, P46410
GLUL
Glutamine synthetase Fusion Protein Ag6309
150 μL
Primary
Human, Pig
Antibody
IgG2b
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1D10G8
Unconjugated
GLUL
GLNS, GLUL, Glutamate ammonia ligase, Glutamate decarboxylase, Glutamine synthetase, GS, PIG43, PIG59
Mouse
Protein A
RUO
2752, 396944
-20°C