missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine Synthetase Mouse anti-Human, Clone: GLUL/6604, Novus Biologicals™
Mouse Monoclonal Antibody
250.00€ - 510.00€
Specifications
| Antigen | Glutamine Synthetase |
|---|---|
| Clone | GLUL/6604 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18367965
|
Novus Biologicals
NBP3-13929-20UG |
20 μg |
250.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18364775
|
Novus Biologicals
NBP3-13929-100UG |
100 μg |
510.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| Glutamine Synthetase | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Hypoxia, Neurodegeneration, Neuroscience, Vision | |
| 10 mM PBS with 0.05% BSA | |
| cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 | |
| Recombinant fragment (around aa 50-250) of human Glutamine Synthetase protein (exact sequence is proprietary) (Uniprot: P15104) | |
| Primary | |
| Store at 4°C. |
| GLUL/6604 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P15104 | |
| 2752 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title