Learn More
Glutamine Synthetase Antibody (CL2999) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46643PCP
Product Code. 30288286
  • 737.00€ / 0.10mL
Description
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glutamine Synthetase
Monoclonal
PerCP
cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
This antibody was developed using a recombinant protein derived from P15104, with the exact immunogen sequence remaining proprietary.
0.1 mL
Hypoxia, Neurodegeneration, Neuroscience, Vision
2752
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL2999
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat