missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut1 Mouse anti-Human, Clone: rGLUT1/2476, Novus Biologicals™
Mouse Monoclonal Antibody
499.00€
Specifications
| Antigen | Glut1 |
|---|---|
| Clone | rGLUT1/2476 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
Glut1 Monoclonal specifically detects Glut1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
| Glut1 | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 6513 | |
| IgG1 κ | |
| Protein A or G purified |
| rGLUT1/2476 | |
| ELISA 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title