missing translation for 'onlineSavingsMsg'
Learn More

GluR7/GRIK3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18448757 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18448757 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18448757 Supplier Novus Biologicals Supplier No. NBP176387AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GluR7/GRIK3 Polyclonal antibody specifically detects GluR7/GRIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR7/GRIK3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias EAA5, Excitatory amino acid receptor 5, GLR7, gluR-7, GluR7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide near the amino terminus of the human Grik3. The immunogen is located within amino acids 30 - 80 of Grik3. Amino Acid Squence: EYADGPNAQVMNA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2899
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.