missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR2 Antibody (7A7D12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61776
This item is not returnable.
View return policy
Description
GluR2 Monoclonal specifically detects GluR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| GluR2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
| Mouse | |
| 99 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 7A7D12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GRIA2 | |
| Purified recombinant fragment of human GRIA2 (AA: 35-175) expressed in E. Coli. | |
| Protein G purified | |
| Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Tumor Suppressors, Vision | |
| 2891 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction