missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose 6 phosphate isomerase Antibody (4F11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43065
This item is not returnable.
View return policy
Description
Glucose 6 phosphate isomerase Monoclonal antibody specifically detects Glucose 6 phosphate isomerase in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Glucose 6 phosphate isomerase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 4F11 | |
| Western Blot 1:2000 - 1:20000, ELISA, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36 | |
| Glucose-6-phosphate isomerase - (Uniprot# P06744) | |
| 100 μg | |
| Cytokine Research, Phospho Specific | |
| 2821 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction