missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose 6 phosphate isomerase Antibody (12A4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26548-50ul
This item is not returnable.
View return policy
Description
Glucose 6 phosphate isomerase Monoclonal antibody specifically detects Glucose 6 phosphate isomerase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| Glucose 6 phosphate isomerase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 12A4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36 | |
| A synthesized peptide derived from Human Glucose 6 phosphate isomerase [UniProt P06744] | |
| 50 μL | |
| Cytokine Research, Phospho Specific | |
| 2821 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction