missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucosamine (N-acetyl)-6-Sulfatase/GNS Antibody, Alexa Fluor™ 647, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Glucosamine (N-acetyl)-6-Sulfatase/GNS Polyclonal specifically detects Glucosamine (N-acetyl)-6-Sulfatase/GNS in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Glucosamine (N-acetyl)-6-Sulfatase/GNS |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.1.6, EC 3.1.6.14, G6Sglucosamine-6-sulfatase, glucosamine (N-acetyl)-6-sulfatase, Glucosamine-6-sulfatase, MGC21274, N-acetylglucosamine-6-sulfatase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Glucosamine (N-acetyl)-6-Sulfatase/GNS (NP_002067.1; Met1-Leu552). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?