missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLTP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10850-1-AP-20UL
This item is not returnable.
View return policy
Description
Accelerates the intermembrane transfer of various glycolipids. Catalyzes the transfer of various glycosphingolipids between membranes but does not catalyze the transfer of phospholipids. May be involved in the intracellular translocation of glucosylceramides.Specifications
| GLTP | |
| Polyclonal | |
| Unconjugated | |
| GLTP | |
| GLTP, glycolipid transfer protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288707, 51228, 56356 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B0BNM9, Q9JL62, Q9NZD2 | |
| Gltp | |
| GLTP Fusion Protein Ag1293 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction