Learn More
Glove Bags
Product Code. 11752134
Change view
Click to view available options
Dimensions (L x W x H):
432 x 432 x 280 mm
432 x 680 x 300 mm
686 x 370 x 457 mm
690 x 690 x 381 mm
940 x 940 x 635 mm
Unit Size:
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11752134 940 x 940 x 635 mm Pack of 6
11786514 686 x 370 x 457 mm Pack of 6
11722134 432 x 432 x 280 mm Pack of 6
11732134 432 x 680 x 300 mm Pack of 6
11742134 690 x 690 x 381 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11752134 Supplier Glas Col Supplier No. 108DX3737H
  • ONLINE EXCLUSIVE PRICE
    878.85€
    390.03€ / Pack of 6
    Save 488.82€
    55% Off
 Restricted Item
Estimated Shipment: 04-12-2026