missing translation for 'onlineSavingsMsg'
Learn More

Norclozapine.HCl, Lipomed

Product Code. 30463620
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30463620

Brand: Lipomed LPMCLZ1882HC10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Norclozapine.HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norclozapine Hydrochloride
CAS 2741767-92-8
Analyte or Component Names Norclozapine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H17 Cl N4 . Cl H
Synonym 5 H-Dibenzo[b,e][1,4 ]diazepine, 8-chloro-11-(1-piperazinyl)-, hydrochloride (1:1) (ACI), 8-Chloro-11-piperazin-1-yl-5 H-dibenzo[b,e][1,4 ]diazepine Hydrochloride, Norclozapine HCl
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.Clc1ccc2Nc3ccccc3C(=Nc2c1)N4CCNCC4
IUPAC Name 3-chloro-6-piperazin-1-yl-11 H-benzo[b][1,4 ]benzodiazepine;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 349.2576
Quantity 10 mg
Formula Weight 348.0909 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.