Learn More
GLI2 Rabbit, anti-Human, Polyclonal, Proteintech
Product Code. 18090586 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18090586 150 μL 150µL
18006675 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18090586 Supplier Proteintech Supplier No. 282451AP150UL
Rabbit Polyclonal Antibody
The GLI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GLI2. This antibody recognizes human antigen. The GLI2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Antigen GLI2
Applications Western Blot, Immunohistochemistry, ELISA
Concentration 400 μg/mL
Gene Symbols GLI family zinc finger 2, GLI2, HPE9, Tax helper protein, THP, THP1, THP2, Zinc finger protein GLI2
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 2736
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.