missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Glass Universal Y-Splitter Capillary Connectors
Description
Advantages compared to stainless steel fittings:
- Provides an easy connection without the need for any tools
- Transparent glass connectors provide optical control during connection
- Features negligible heat capacity and no dead volume
Specifications
Specifications
| Type | Connector |
| Color | Clear |
| Packaging Quantity | 1 Each |
| Description | Slightly tapered inner diameter |
| For Use With | 3 Columns of 0.20 to 0.53 mm I.D. |
| Material | Glass |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?