Learn More
Trajan Scientific and Medical™ Glass Lined Tubing
Click to view available options
Overall Length (Metric):
30 cm
60 cm
Diameter (Metric) Inner:
0.3 mm
0.5 mm
1.8 mm
4 mm
Diameter (Metric) Outer:
1.59 mm
3.175 mm
6.35 mm
Unit Size:
300mm
Each
Description
- Excellent reistance to strong acids and bases
- Ideal for HPLC columns - particularly for biological matrices
- Temperature stable up to 500°C
Specifications
Specifications
| Type | Tubing |
| Diameter (English) Outer | 1/16 in. |
| Diameter (Metric) Inner | 0.3 mm |
| Diameter (Metric) Outer | 1.59 mm |
| Overall Length (Metric) | 30 cm |
| Material | Stainless Steel, Borosilicate Glass (Lining) |
| Max. Operating Temperature (Metric) | 500°C |