Learn More
Lenz Laborglasintrumente Glass Filter Plugs with Tube
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07209425
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10048269
  • 78.44€ / Each
Estimated Shipment: 28-08-2024
Description
  • Filter plugs
  • Cylindrical
  • With tube (dia. 8 mm x length 210 mm)
  • Material: borosilicate glass 3.3
Specifications
Specifications
Borosilicate Glass (3.3)
4
25 mm