Learn More
Lenz Laborglasintrumente Glass Filter Plugs with Tube
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
63.23€ - 78.44€
Product Code | Brand | Porosity | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Diameter (Metric) | Price | Quantity & Availability | ||||
10048269
|
Lenz Laborglasintrumente
07209425 |
4 |
78.44€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11516784
|
Lenz Laborglasintrumente
07208020 |
0 | 9 mm |
63.23€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11526784
|
Lenz Laborglasintrumente
07208120 |
1 | 9 mm |
63.23€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11536784
|
Lenz Laborglasintrumente
07208220 |
2 | 9 mm |
63.23€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11546784
|
Lenz Laborglasintrumente
07208320 |
3 | 9 mm |
63.23€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11556784
|
Lenz Laborglasintrumente
07208420 |
4 | 9 mm |
70.86€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11566784
|
Lenz Laborglasintrumente
07209025 |
0 | 13 mm |
64.13€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11576784
|
Lenz Laborglasintrumente
07209125 |
1 | 13 mm |
64.13€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11586784
|
Lenz Laborglasintrumente
07209225 |
2 | 13 mm |
64.13€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11596784
|
Lenz Laborglasintrumente
07209325 |
3 | 13 mm |
64.13€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
Description
- Filter plugs
- Cylindrical
- With tube (dia. 8 mm x length 210 mm)
- Material: borosilicate glass 3.3
Specifications
Borosilicate Glass (3.3) |