missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GKN1, Mouse anti-Human, Clone: 1G17, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00056287-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: NYNINVNDDNNNAGSGQQSVSVNNEHNVANVDNNNGWDSWNSIWDYGNGFAATRLFQKKTCIVHKMNKEVMPSIQSLDALVKEKKLQGKGPGGPPPKGLMYSVNPNKVDDLSKFGKNIANMCRGIPTYMAEEMQEASLFFYSGTCYTTSVLWIVDISFCGDTVENSpecifications
| GKN1 | |
| Monoclonal | |
| Unconjugated | |
| gastrokine 1 | |
| AMP18/BRICD1/CA11/FOV/MGC70354/foveolin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1G17 | |
| In 1x PBS, pH 7.4 | |
| BC059778 | |
| GKN1 | |
| GKN1 (AAH59778, 21 a.a. ∼ 185 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 56287 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction