missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR Ligand/TNFSF18 Antibody (994529), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB21773P
490.45 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOF
Specifications
| GITR Ligand/TNFSF18 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Neutralization, CyTOF | |
| 994529 | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18, Thr47-Ser173, Accession # Q7TS55 | |
| 0.1 mL | |
| Apoptosis | |
| 8995 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction