missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR Ligand Rat anti-Mouse, BUV496, Clone: MIH44, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 750026
This item is not returnable.
View return policy
Description
GITR Ligand Monoclonal antibody specifically detects GITR Ligand in Mouse samples. It is validated for Flow Cytometry
Specifications
| GITR Ligand | |
| Monoclonal | |
| Brilliant Ultraviolet 496 | |
| Tnfsf18; Gitrl Glucocorticoid-induced-tumor necrosis factor receptor ligand | |
| Mouse GITRL Transfected Cell Line | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| MIH44 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction