missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIT1 Antibody (S39B-8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
174.00€
Specifications
| Antigen | GIT1 |
|---|---|
| Clone | S39B-8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
Description
GIT1 Monoclonal specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Knockout Validated.Specifications
| GIT1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 | |
| GIT1 | |
| IgG1 | |
| Protein G purified | |
| Detects approx 90kDa. Does not cross-react with GIT2. |
| S39B-8 | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9Z272 | |
| 28964 | |
| Fusion protein amino acids 375-770 (C-terminus) of rat GIT1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title