missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GIRK2 Monoclonal antibody specifically detects GIRK2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | GIRK2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | KCNJ6/7559 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | BIR1Kir3.2, G protein-activated inward rectifier potassium channel 2, GIRK-2, GIRK2KATP-2, hiGIRK2, Inward rectifier K(+) channel Kir3.2, inward rectifier potassium channel KIR3.2, KATP2human G protein coupled inward rectifier potassium channel 2 (hiGIRK2)10KCNJ7KIR3.2, MGC126596, Potassium channel, inwardly rectifying subfamily J member 6, potassium inwardly-rectifying channel, subfamily J, member 6 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?