missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF128 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149160
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
GRAIL is a type I transmembrane protein that localizes to the endocytic pathway and contains a PA (protease associated) domain and a RING-type zinc finger domain. It is a ubiquitin-protein isopeptide ligase (E3) necessary for the induction of CD4(+) T cell anergy in vivo and induced expression of this protein was observed in anergic CD4 (+) T cells, which suggested a role in the induction of anergic phenotype. It is differentially expressed in naturally occurring and peripherally induced CD25 (+) T regulatory cells and the expression is linked to the conversion of these cells to a regulatory phenotype. Expression of GRAIL in retrovirally transduced T cell hybridomas limits activation-induced IL-2 and IL-4 cytokine production. It also functions in the patterning of the dorsal ectoderm and sensitizes ectoderm to respond to neural-inducing signals.
Specifications
| RNF128 | |
| Polyclonal | |
| Unconjugated | |
| RNF128 | |
| 1300002C13Rik; AI987883; E3 ubiquitin-protein ligase RNF128; Gene related to anergy in lymphocytes protein; goliath-related E3 ubiquitin-protein ligase 1; Grail; Greul1; MNCb-3816; RGD1566282; RING finger protein 128; ring finger protein 128, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF128; RNF128; RP11-150F24.1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 79589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8TEB7 | |
| RNF128 | |
| E. coli-derived recombinant human GRAIL/RNF128, Val42-Pro161. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction