missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | GIPC1 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231697
|
Novus Biologicals
NBP3-35151-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226466
|
Novus Biologicals
NBP3-35151-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GIPC1 Polyclonal antibody specifically detects GIPC1 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| GIPC1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 10755 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C19orf3chromosome 19 open reading frame 3, GAIP C-terminus-interacting protein, GIPC PDZ domain containing family, member 1, GIPCRGS-GAIP-interacting protein, GLUT1 C-terminal binding protein, GLUT1CBP, IGF-1 receptor interacting protein 1, MGC15889, MGC3774, NIP, PDZ domain-containing protein GIPC1, regulator of G-protein signalling 19 interacting protein 1, RGS19IP1IIP-1, SEMCAP, SYNECTIIN, SYNECTIN, Tax interaction protein 2, TIP-2RGS19-interacting protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-333 of human GIPC1 (NP_005707.1).,, Sequence:, MPLGLGRRKKAPPLVENEEAEPGRGGLGVGEPGPLGGGGSGGPQMGLPPPPPALRPRLVFHTQLAHGSPTGRIEGFTNVKELYGKIAEAFRLPTAEVMFCTLNTHKVDMDKLLGGQIGLEDFIFAHVKGQRKEVEVFKSEDALGLTITDNGAGYAFIKRIKEGSVIDHIHLISVGDMIEAINGQSLLGCRHYEVARLLKELPRGRTFTLKLTEPRKAFDMISQRSAGGRPGSGPQLGTGRGTLRLRSRGPATVEDLPSAFEEKAIEKVDDLLESYMGIRDTELAATMVELGKDKRNPDELAEALDERLGDFAFPDEFVFDVWGAIGDAKVGRY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title