missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIMAP6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 595.00€
Specifications
| Antigen | GIMAP6 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618740
|
Novus Biologicals
NBP2-76542-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18326751
|
Novus Biologicals
NBP2-76542 |
100 μL |
595.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GIMAP6 Polyclonal specifically detects GIMAP6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| GIMAP6 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZp686A01175, FLJ22690, GTPase IMAP family member 6, GTPase, IMAP family member 6, hIAN2, hIAN6, human immune associated nucleotide 2, IAN2, IAN-2, IAN6, IAN-6, immune associated nucleotide 2, immune associated nucleotide 6, Immunity-associated nucleotide 2 protein, Immunity-associated nucleotide 6 protein | |
| GIMAP6 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Cancer | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 474344.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SLEDYVRETNNQALAWLDVTLARRHCGFNNRAQGEEQEAQLRELMEKVEAIMWENEGDYYSNKAYQYTQQNFRLKELQERQVSQGQGSEDVPGEESWLEGLSQIQK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title