missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIMAP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13445-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detected one 28 kDa band in several tissues.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.Specifications
| GIMAP2 | |
| Polyclonal | |
| Unconjugated | |
| GIMAP2 | |
| GIMAP2, GTPase IMAP family member 2, GTPase, IMAP family member 2, HIMAP2, IMAP2, Immunity associated protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26157 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UG22 | |
| GIMAP2 | |
| GIMAP2 Fusion Protein Ag4273 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction