Learn More
Gilson™ PIPETMANTM DIAMOND Reload Pack Empty Box Set
Product Code. 15279386 Shop All Gilson Products
Change view
Click to view available options
Holds:
D10 Pipette Tips (0.1 to 10μL), D200 Pipette Tips (2 to 200μL)
D1000 Pipette Tips (100 to 1000μL)
Array:
10 x 96
6 x 96
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Array Unit Size
15279386 D10 Pipette Tips (0.1 to 10μL), D200 Pipette Tips (2 to 200μL) 10 x 96 Pack of 2
15209396 D1000 Pipette Tips (100 to 1000μL) 6 x 96 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15279386 Supplier Gilson™ Supplier No. F171204
  • ONLINE EXCLUSIVE PRICE
    16.40€
    15.75€ / Pack of 2
    Save 0.65€
    3% Off
In Stock
Add to basket
Add to basket
Bulk Quote Request
PIPETMANTM Diamond Reload Pack Empty Box Set for blister refill and reload packs. Easy to store on all benches.
  • Hold tips of model DL10 (0.1 to 20 μL), D200 (2 to 200 μL), D1000 (100 to 1000 μL)
  • 70% less plastic
  • Three times smaller than regular tips box
  • Eco-friendly
  • Available in set of 2
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Clear
Holds D10 Pipette Tips (0.1 to 10μL), D200 Pipette Tips (2 to 200μL)
Material Polypropylene
Positions 1
Shape Rectangular
Stackable Yes, Interlocking
Sterility Non-sterile
For Use With (Equipment) Diamond Tips
Quantity Set of 2
Size 134 x 89 x 127 mm
Length (Metric) 9.3 cm
Length (English) 3.66 in.
Width (English) 5.28 in.
Height (English) 4.92 in.
Includes 2 Empty Racks
Product Type Empty Box Reload
Array 10 x 96