missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFRA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25379-1-AP-150UL
This item is not returnable.
View return policy
Description
GFRa-3 binds persephin, thus, persephin, GFRa-3, and Ret PTK form a complex to transduce persephin signal and to mediate persephin function.Specifications
| GFRA3 | |
| Polyclonal | |
| Unconjugated | |
| GFRA3 | |
| GDNF family receptor alpha 3, GDNF receptor alpha 3, GDNFR alpha 3, GFR alpha 3, GFRA3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14587, 2676 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35118, O60609 | |
| Gfra3 | |
| GFRA3 Fusion Protein Ag13253 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction