missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFR alpha-3/GDNF R alpha-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99078-100ul
This item is not returnable.
View return policy
Description
GFR alpha-3/GDNF R alpha-3 Polyclonal antibody specifically detects GFR alpha-3/GDNF R alpha-3 in Human samples. It is validated for ELISA
Specifications
| GFR alpha-3/GDNF R alpha-3 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| GDNF family receptor alpha 3, GDNF family receptor alpha-3, GDNF receptor alpha-3, GDNFR-alpha-3, GFRa-3, GFR-alpha-3, glial cell line-derived neurotrophic factor receptor alpha-3, GPI-linked receptor | |
| Produced in rabbits immunized with purified, recombinant Human GFR alpha-3/GDNF R alpha-3 (Accession#: NP_001487.2; Met 1-Trp 382) | |
| 100 μg | |
| Cytokine Research, Immunology | |
| 2676 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction